missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium metaborate, 99.992% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01298814

 View more versions of this product

Catalog No. AA1298814



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Calcium metaborate
13701-64-9
25 g
B2CaO4
MFCD00049397
ILOKQJWLMPPMQU-UHFFFAOYSA-N
calcium;oxido(oxo)borane
10290800
99.992%
White
Powder
(metals basis)
Ca(BO2)2
calcium metaborate, 2bo2.ca, calcium dioxoborinate, dimetaboric acid calcium salt, calcium 2+ diboron oxide hydroxide
B(=O)[O-].B(=O)[O-].[Ca+2]
125.694
125.7
Odorless

Chemical Identifiers

13701-64-9
125.694
ILOKQJWLMPPMQU-UHFFFAOYSA-N
10290800
B(=O)[O-].B(=O)[O-].[Ca+2]
B2CaO4
MFCD00049397
calcium metaborate, 2bo2.ca, calcium dioxoborinate, dimetaboric acid calcium salt, calcium 2+ diboron oxide hydroxide
calcium;oxido(oxo)borane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-224-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only