missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium L-threonate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6627936

 View more versions of this product

Catalog No. AAJ6627936



Description

Description

Calcium L-threonate is reactant involved in synthesis of cyclic hydroxamates and substituted oxazinanones and oxidative degradation to yield oxalate complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium L-threonate is reactant involved in synthesis of cyclic hydroxamates and substituted oxazinanones and oxidative degradation to yield oxalate complexes.

Solubility
Soluble in water. Sparingly soluble in methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Calcium L-threonate
>300°C
C8H14CaO10
500 g
Soluble in water. Sparingly soluble in methanol.
[Ca++].OCC(O)C(O)C([O-])=O.OCC(O)C(O)C([O-])=O
310.27
310.27
Powder
70753-61-6
Brown
MFCD00077882,MFCD00150824
calcium l-threonate, l-threonic acid calcium salt, calcium 2r,3s-2,3,4-trihydroxybutanoate, calcium threonate, unii-hbb4ypo93u, hbb4ypo93u, calcium bis l-threonate, threonine, calcium salt, bis l-threonic acid calcium salt, butanoic acid, 2,3,4-trihydroxy-, calcium salt 2:1 , 2r,3s
ZJXGOFZGZFVRHK-UHFFFAOYNA-L
calcium;(2R,3S)-2,3,4-trihydroxybutanoate
13388558
98%

Chemical Identifiers

70753-61-6
310.27
ZJXGOFZGZFVRHK-UHFFFAOYNA-L
13388558
[Ca++].OCC(O)C(O)C([O-])=O.OCC(O)C(O)C([O-])=O
C8H14CaO10
MFCD00077882,MFCD00150824
calcium l-threonate, l-threonic acid calcium salt, calcium 2r,3s-2,3,4-trihydroxybutanoate, calcium threonate, unii-hbb4ypo93u, hbb4ypo93u, calcium bis l-threonate, threonine, calcium salt, bis l-threonic acid calcium salt, butanoic acid, 2,3,4-trihydroxy-, calcium salt 2:1 , 2r,3s
calcium;(2R,3S)-2,3,4-trihydroxybutanoate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only