Please login to your online account to display your discounted pricing

Calcium Ionophore A23187 (Free Acid), Fisher BioReagents

Isolated from Streptomyces chartreusensis

$140.66 - $449.59

Chemical Identifiers

Chemical Name or Material Calcium Ionophore A23187
CAS 52665-69-7
Physical Form Solid
Molecular Formula C29H37N3O6
Formula Weight 523.6
Synonym Calcimycin
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP595-5

  SDS
fisher scientific company
FLBP5955
Poly Micro Tube 5mg Each for $247.41

BP595-1

  SDS
fisher scientific company
FLBP5951
Poly Micro Tube 1mg Each for $140.66
Case of 6 Each for $843.96

BP595-10

  SDS
fisher scientific company
FLBP59510
Poly Micro Tube 10mg Each for $449.59
Description & Specifications

Specifications

Formula Weight 523.6
Melting Point 182°C
Quantity 5mg
Merck Index 15, 1642
Physical Form Solid
Molecular Formula C29H37N3O6
Color Off-white
Chemical Name or Material Calcium Ionophore A23187
CAS 52665-69-7
Health Hazard 1 Signal Word:WARNING!
Packaging Poly Micro Tube
Carbon (C) 65.52 to 67.52%
Identification Pass Test
Hydrogen (H) 6.92 to 7.32%
Synonym Calcimycin
Nitrogen (N) 7.82 to 8.22%
Purity 98% min.
Specific Rotation -50 to -56° (+ or -)
TLC ≥98% (purity)
Recommended Storage 4°C
ChemAlert Storage Symbol Gray
Health Hazard 3 Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Irritating to eyes, respiratory system and skin.
NFPA
Health:

This widely used ionophore increases the permeability of biological membranes for bivalent cations.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Signal Word:WARNING!

Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Irritating to eyes, respiratory system and skin.
NFPA
Health:

4°C