Please login to your online account to display your discounted pricing

Calcium Ionophore A23187 (Free Acid), Fisher BioReagents


Isolated from Streptomyces chartreusensis

Quantity 1mg
Packaging Poly Micro Tube

Manufacturer: fisher scientific company  FLBP5951

 View more versions of this product

Catalog No. BP595-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 523.6
Melting Point 182°C
Quantity 1mg
Physical Form Solid
Molecular Formula C29H37N3O6
Color Off-white
Merck Index 15, 1642
Carbon (C) 65.52 to 67.52%
Identification Pass Test
CAS 52665-69-7
Synonym Calcimycin
Packaging Poly Micro Tube
Purity 98% min.
Specific Rotation -50 to -56° (+ or -)
TLC ≥98% (purity)
Chemical Name or Material Calcium Ionophore A23187
Hydrogen (H) 6.92 to 7.32%
ChemAlert Storage Symbol Gray
Nitrogen (N) 7.82 to 8.22%
Health Hazard 1 Signal Word:WARNING!
Health Hazard 3 Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Irritating to eyes, respiratory system and skin.
NFPA
Health:
Recommended Storage 4°C

Specifications

Chemical Name or Material Calcium Ionophore A23187
CAS 52665-69-7
Physical Form Solid
Molecular Formula C29H37N3O6
Formula Weight 523.6
Synonym Calcimycin
Accessories
 

Signal Word:WARNING!

Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Irritating to eyes, respiratory system and skin.
NFPA
Health:

4°C

SDS & Safety Documents