Learn More
Calcium iodide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04712114
Description
It is used in photography. It is also used in cat food as a source of iodine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
1100°C | |
784°C | |
10102-68-8 | |
Beads | |
(metals basis) | |
MFCD00010910 | |
calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g | |
UNMYWSMUMWPJLR-UHFFFAOYSA-L | |
calcium;diiodide | |
5462722 | |
293.89 | |
Odorless | |
Hygroscopic; Light sensitive |
Calcium iodide | |
Ultra dry | |
White to Yellow | |
25 g | |
CaI2 | |
14,1677 | |
Soluble in water (64.6g/100 mL (0°C)). Soluble in acetone and alcohols. | |
[Ca+2].[I-].[I-] | |
293.887 | |
CHEBI:81718 | |
99.995% | |
Ampule | |
3.956 g/mL |
Chemical Identifiers
10102-68-8 | |
293.887 | |
UNMYWSMUMWPJLR-UHFFFAOYSA-L | |
5462722 | |
calcium;diiodide |
CaI2 | |
MFCD00010910 | |
calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g | |
CHEBI:81718 | |
[Ca+2].[I-].[I-] |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 233-276-8
TSCA : Yes
storageNote1 :
- Ampouled under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only