missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium iodide, ultra dry, 99.99% (metals basis), Alfa Aesar™

Light Sensitive and Hygroscopic

Manufacturer:  Alfa Aesar 4367422

 View more versions of this product

Catalog No. AA4367422



Specifications

Specifications

Calcium iodide
99.99% (Metal basis)
MFCD00010910
acmc-1c61l
[Ca+2].[I-].[I-]
293.887
CHEBI:81718
Powder
100g
10102-68-8
CaI2
Off-white to pale yellow powder or beads
UNMYWSMUMWPJLR-UHFFFAOYSA-L
calcium;diiodide
5462722
293.89
White to Yellow

Chemical Identifiers

10102-68-8
293.887
UNMYWSMUMWPJLR-UHFFFAOYSA-L
5462722
calcium;diiodide
CaI2
MFCD00010910
acmc-1c61l
CHEBI:81718
[Ca+2].[I-].[I-]
Safety and Handling

Safety and Handling

H315-H319 : Causes skin irritation. Causes serious eye irritation.

EINECSNumber : 233-276-8

Documents