Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium iodide hydrate, Puratronic™, 99.9965% (metals basis)

Chemical Identifiers

Chemical Name or Material Calcium iodide hydrate
CAS 71626-98-7
Purity Grade Puratronic™
Formula Weight 293.89 (Anhydrous)
MDL Number MFCD00149622
Merck Index 14,1677
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1111714

  SDS
alfa aesar™
1111714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1111706

  SDS
alfa aesar™
1111706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 293.89 (Anhydrous)
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Chemical Name or Material Calcium iodide hydrate
TSCA TSCA
Purity Grade Puratronic™
Physical Form Crystalline
Assay 99.9965%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00149622
CAS 71626-98-7
Merck Index 14,1677
Color Yellow
Melting Point 42°C (dec.)
Quantity 25g
Solubility Information Freely soluble in water and alcohol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Density 2.55g/mL
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.