Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium iodide hydrate

Chemical Identifiers

Chemical Name or Material Calcium iodide hydrate
CAS 71626-98-7
Formula Weight 293.89 (Anhydrous)
MDL Number MFCD00149622
Merck Index 14,1677
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4013622

  SDS
alfa aesar™
4013622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4013636

  SDS
alfa aesar™
4013636
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Lumps
Formula Weight 293.89 (Anhydrous)
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Chemical Name or Material Calcium iodide hydrate
TSCA TSCA
Sensitivity Hygroscopic
MDL Number MFCD00149622
CAS 71626-98-7
Merck Index 14,1677
Color White to gray
Melting Point 42°C (dec.)
Quantity 100g
Solubility Information Freely soluble in water and alcohol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Density 2.55g/mL
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.