missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium iodide, anhydrous, 99.5% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 2239514

 View more versions of this product

Catalog No. AA2239514



Specifications

Specifications

Calcium iodide
10102-68-8
(metals basis)
MFCD00010910
calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g
UNMYWSMUMWPJLR-UHFFFAOYSA-L
calcium;diiodide
5462722
293.89
99.5%
−20 Mesh
Light Sensitive & Hygroscopic
White to Yellow
25g
Anhydrous
99.5% (Metals basis)
CaI2
14,1677
Freely soluble in water, methanol, ethanol, acetone; Insoluble in ether
[Ca+2].[I-].[I-]
293.887
CHEBI:81718
Powder
3.956g/mL
Odorless
1100°C
784°C

Chemical Identifiers

10102-68-8
293.887
UNMYWSMUMWPJLR-UHFFFAOYSA-L
5462722
calcium;diiodide
CaI2
MFCD00010910
calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g
CHEBI:81718
[Ca+2].[I-].[I-]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA

SDS Documents