Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium iodide, anhydrous, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Calcium iodide
CAS 10102-68-8
Name Note Anhydrous
Molecular Formula CaI2
Formula Weight 293.89
MDL Number MFCD00010910
Merck Index 14,1677
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2239514

  SDS
alfa aesar™
2239514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2239522

  SDS
alfa aesar
2239522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 293.89
Assay Percent Range 99.5% (Metals basis)
Molecular Formula CaI2
Chemical Name or Material Calcium iodide
TSCA TSCA
Mesh Size −20 Mesh
Odor Odorless
Name Note Anhydrous
Physical Form Powder
Assay 99.5%
Sensitivity Light Sensitive & Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00010910
CAS 10102-68-8
Merck Index 14,1677
Boiling Point 1100°C
Color White to pale yellow
Melting Point 784°C
Quantity 25g
Solubility Information Freely soluble in water, methanol, ethanol, acetone; Insoluble in ether
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Density 3.956g/mL
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.