Please login to your online account to display your discounted pricing

Calcium hypochlorite, tech., granular, 65% available chlorine, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  300341000

 View more versions of this product

Catalog No. AC300341000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥65% (available Cl)
pH ≈11.6 (1% aq. soln.)
Formula Weight 142.98
Purity Grade Technical Grade
Physical Form Granular Solid or Granules
Quantity 100g
Color White to off-white or faint green
EINECS Number 231-908-7
TSCA TSCA
RTECS Number NH3485000
Water 12.50%
Sodium Chloride 8.5%
Calcium Hydroxide 4.5%
Calcium Chlorate 1.0%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
May intensify fire; oxidizer.
Contact with acids liberates toxic gas.
Health Hazard 1 Danger
Chemical Name or Material Calcium hypochlorite
CAS 7778-54-3
Molecular Formula CaCl2O2
Linear Formula Ca(OCl)2
Name Note tech.
MDL Number MFCD00010900
Calcium Chloride 1.0%
Available Chlorine 72%
Packaging Plastic bottle
Additional Information others: 0.5%

Specifications

Chemical Name or Material Calcium hypochlorite
Name Note tech.
CAS 7778-54-3
Purity Grade Technical Grade
Assay ≥65% (available Cl)
Molecular Formula CaCl2O2
Linear Formula Ca(OCl)2
Formula Weight 142.98
MDL Number MFCD00010900
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
May intensify fire; oxidizer.
Contact with acids liberates toxic gas.

SDS & Safety Documents