missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium hypochlorite, tech., granular, 65% available chlorine, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC300340050

 View more versions of this product

Catalog No. AC300340050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Calcium hypochlorite
7778-54-3
Technical Grade
≥65% (available Cl)
Ca(OCl)2
MFCD00010900
4.5%
1.0%
8.5%
White to off-white or faint green
5kg
tech.
others: 0.5%
Granular Solid or Granules
CaCl2O2
142.98
72%
1.0%
Plastic drum
12.50%
≈11.6 (1% aq. soln.)

Chemical Identifiers

7778-54-3
Ca(OCl)2
MFCD00010900
≥65% (available Cl)
CaCl2O2
142.98
Calcium hypochlorite, tech.
Technical Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
May intensify fire; oxidizer.
Contact with acids liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 231-908-7

RTECSNumber : NH3485000

TSCA : TSCA