Please login to your online account to display your discounted pricing

Calcium Hypochlorite, Purified, 65% minimum Available Chlorine, Granular, Purified, J.T.Baker™

$327.19 - $327.19

Chemical Identifiers

Chemical Name or Material Calcium Hypochlorite
CAS 7778-54-3
Name Note Purified
Purity Grade Purified
Molecular Formula CaCl2O2
Formula Weight 142.98
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002534

 
avantor performance
137805
Poly Bottle 2.5kg Each for $327.19
Description & Specifications

Specifications

Assay ≥65% available chlorine
Formula Weight 142.98
Chemical Name or Material Calcium Hypochlorite
Name Note Purified
Physical Form Granular
Specific Gravity 1L = 2.35kg
Molecular Formula CaCl2O2
CAS 7778-54-3
Packaging Poly Bottle
Purity Grade Purified
Quantity 2.5kg