Please login to your online account to display your discounted pricing

Calcium Hypochlorite (Granular/Certified), Fisher Chemical

$286.15 - $286.15

Chemical Identifiers

Chemical Name or Material Calcium Hypochlorite
CAS 7778-54-3
Purity Grade Certified Grade
Physical Form Solid
Molecular Formula Ca Cl2O2
Linear Formula Ca(OCl)2
Formula Weight 142.98
MDL Number MFCD00010900
Synonym Calcium Oxychloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

C100-500

  SDS
fisher scientific company
FLC100500
Glass Bottle 500g Each for $286.15
Case of 6 Each for $1,716.90
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 142.98
Melting Point 100°C
Quantity 500g
DOT Information DOT Class 5.1, : Oxidizer
Physical Form Solid
Molecular Formula Ca Cl2O2
Color Off-white
pH 11.4
Density 2.350g/cm³
Chemical Name or Material Calcium Hypochlorite
Identification Pass Test
MDL Number MFCD00010900
Synonym Calcium Oxychloride
Available Chlorine 65% min.
Iron (Fe) 0.5% max.
Packaging Glass Bottle
Aluminum Oxide 0.1% max.
Sulfate 0.05% max.
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Harmful if swallowed. Contact with acids liberates toxic gas. Very toxic to aquatic organisms. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:0
Instability:1
Linear Formula Ca(OCl)2
CAS 7778-54-3
ChemAlert Storage Symbol Yellow

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Harmful if swallowed. Contact with acids liberates toxic gas. Very toxic to aquatic organisms. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:0
Instability:1

DANGER!