Please login to your online account to display your discounted pricing

Calcium Hydroxide, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$143.36 - $219.62

Chemical Identifiers

Chemical Name or Material Calcium Hydroxide
CAS 1305-62-0
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Ca(OH)2
Formula Weight 74.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002532

 
avantor performance
137201
Poly Bottle 500g Each for $143.36

02002531

 
avantor performance
137205
Poly Pail 2kg Each for $219.62
Description & Specifications

Specifications

Formula Weight 74.09
Purity Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Calcium Hydroxide
Physical Form Powder
Specific Gravity 1L = 2.24kg
Molecular Formula Ca(OH)2
CAS 1305-62-0
Packaging Poly Bottle
Quantity 500g