missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, Powder, ACS, 95%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C111512KG

 View more versions of this product

Catalog No. 18609488


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1305-62-0
12 kg
MFCD00010901
[OH-].[OH-].[Ca++]
74.09
ACS
1
CaH2O2
AXCZMVOFGPJBDE-UHFFFAOYSA-L
calcium dihydroxide
95%
Poly Pail

Chemical Identifiers

1305-62-0
74.09
AXCZMVOFGPJBDE-UHFFFAOYSA-L
[OH-].[OH-].[Ca++]
CaH2O2
MFCD00010901
calcium dihydroxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.