Please login to your online account to display your discounted pricing

Calcium Hydroxide ACS AR Powder, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Calcium Hydroxide
CAS 1305-62-0
Purity Grade ACS Grade
Physical Form Powder
Molecular Formula Ca(OH)2
Formula Weight 74.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK4195212

 
avantor performance
419505
STAKMOR 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 74.09
Purity Grade ACS Grade
Quantity 2.5kg
Molecular Formula Ca(OH)2
Physical Form Powder
Packaging STAKMOR
CAS 1305-62-0
Chemical Name or Material Calcium Hydroxide