missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium hydroxide, 99.995%, (trace metal basis), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 385610050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for ultra trace analysisSpecifications
Calcium hydroxide | |
5g | |
H2CaO2 | |
15,1675 | |
Solubility in water: 1.65g/L (20°C). Other solubilities: soluble in acids (evolution of heat) and nh4cl sol,soluble in glycerol and sugar,insoluble in alcohol | |
[OH-].[OH-].[Ca+2] | |
74.09 | |
CHEBI:31341 | |
99.995% | |
Glass bottle |
1305-62-0 | |
54% min. (Ca) (Complexometry) | |
Ca(OH)2 | |
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid | |
AXCZMVOFGPJBDE-UHFFFAOYSA-L | |
calcium;dihydroxide | |
6093208 | |
74.09 | |
Trace Metal Basis | |
2.24g/mL |
Chemical Identifiers
1305-62-0 | |
74.09 | |
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid | |
CHEBI:31341 | |
[OH-].[OH-].[Ca+2] |
H2CaO2 | |
AXCZMVOFGPJBDE-UHFFFAOYSA-L | |
6093208 | |
calcium;dihydroxide |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 215-137-3
RUO â Research Use Only