Please login to your online account to display your discounted pricing

Calcium hydroxide, 98%, extra pure, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  219181000

 View more versions of this product

Catalog No. AC219181000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Purity Grade Extra Pure
Physical Form Amorphous Powder
Formula Weight 74.09
Quantity 100g
Color Off-white
Merck Index 15, 1675
Density 2.24g/mL
Heavy Metals (as Pb) 40ppm max.
Insoluble Matter 0.2% max. (in HCl)
Linear Formula Ca(OH)2
Chemical Name or Material Calcium hydroxide
Molecular Formula H2CaO2
MDL Number MFCD00010901
Identification Passes Tests
Packaging Plastic bottle
CAS 1305-62-0
Arsenic (As) 3ppm max.
Additional Information Mg and alkali salts (SO4): < 2.5%
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.

Specifications

Chemical Name or Material Calcium hydroxide
CAS 1305-62-0
Purity Grade Extra Pure
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula H2CaO2
Linear Formula Ca(OH)2
Formula Weight 74.09
MDL Number MFCD00010901
Accessories

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.