missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium hydride, ca. 93%, extra pure, 10-100 mm pieces, ACROS Organics™

Manufacturer:  Acros Organics 199961000

 View more versions of this product

Catalog No. AC199961000



Description

Description

Adsorbent, Reduction reagent
Specifications

Specifications

Calcium hydride
7789-78-8
ca. 93%
CaH2
42.09
ca. 93%
5000ppm max. Typically
980mL/g min. (Typically) CaH2
8000ppm max. Typically
Plastic bottle
1.9
675.0°C
ca. 93%
100ppm max. Typically
CaH2
MFCD00010897
Irregular Lumps or Pieces
Extra Pure
1.9000g/mL
100ppm max. Typically
2000ppm max. Typically
Solubility in water: reacts. Other solubilities: decomposes with lower alcohols and carboxylic acid, soluble only in salt melts
Gray
100g

Chemical Identifiers

7789-78-8
CaH2
MFCD00010897
Safety and Handling

Safety and Handling

GHS H Statement:
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Handle under inert gas. Protect from moisture.
Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages.
Store in a dry place. Store in a closed container.
In case of fire: Use dry sand, dry chemical or alcohol-resistant foam for extinction.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 232-189-2

TSCA : TSCA

SDS Documents