missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium hydride, ca. 93%, extra pure, 1-20 mm grain size, up to 15% powder, Acros Organics

$142.56 - $480.62

Chemical Identifiers

CAS 7789-78-8
Molecular Formula CaH2
Molecular Weight (g/mol) 42.09
MDL Number MFCD00010897
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC196791000 SDS View Documents Fisher Scientific Acros
196791000
100g Plastic bottle Each for $142.56
 
AC196795000 SDS View Documents Fisher Scientific Acros
196795000
500g Plastic bottle Each for $480.62
 
Description

Description

Adsorbent, Reduction reagent
Specifications

Chemical Identifiers

7789-78-8
CaH2
42.09
MFCD00010897

Specifications

7789-78-8
92% min. (Total Ca)
CaH2
1404
15, 1674
42.09
Granular Powder
Extra Pure
5000ppm max.
> 980mL/g CaH2
8000ppm max.
Plastic bottle
1.9
675.0°C
Calcium hydride, ca. 93%
100ppm max.
CaH2
MFCD00010897
01,105
Solubility in water: reacts. Other solubilities: decomposes with lower alcohols and carboxylic acid,soluble only in salt melts
42.09
ca. 93%
5000ppm max
1.9000g/mL
100ppm max.
2000ppm max.
Solubility in water: reacts. Other solubilities: decomposes with lower alcohols and carboxylic acid, soluble only in salt melts
Gray
100g
Safety and Handling

Safety and Handling

GHS H Statement
In contact with water releases flammable gases which may ignite spontaneously.
Causes skin irritation.
Causes serious eye irritation.
Reacts violently with water.

GHS P Statement
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Brush off loose particles from skin.
Immerse in cool water/wrap in wet bandages.
IF ON S

GHS Signal Word: Danger

EINECSNumber : 232-189-2

TSCA : TSCA