Please login to your online account to display your discounted pricing

Calcium hydride, 90-95%, Alfa Aesar™

$456.55 - $456.55

Chemical Identifiers

Chemical Name or Material Calcium hydride
CAS 7789-78-8
Molecular Formula CaH2
Formula Weight 42.1
MDL Number MFCD00010897
UN Number UN1404
Merck Index 14,1672
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1910636

  SDS
alfa aesar
1910636*
Each for $456.55
Description & Specifications

Specifications

Physical Form 2mm and down
Formula Weight 42.1
Quantity 500g
Chemical Name or Material Calcium hydride
TSCA TSCA
Odor Odorless
Assay 90 to 95%
Sensitivity Moisture Sensitive
Molecular Formula CaH2
MDL Number MFCD00010897
CAS 7789-78-8
UN Number UN1404
Merck Index 14,1672
Melting Point 600°C (dec.)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H260;
In contact with water releases flammable gases which may ignite spontaneously.
Health Hazard 3 GHS P Statement
P231+P232-P280-P233-P370+P378c-P402+P404-P501a;
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 1.900
DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: CALCIUM HYDRIDE
Safety and Handling

Danger

GHS P Statement
P231+P232-P280-P233-P370+P378c-P402+P404-P501a;
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H260;
In contact with water releases flammable gases which may ignite spontaneously.