missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium hydride, 90-95%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1910636*

 View more versions of this product

Catalog No. AA1910636



Specifications

Specifications

Calcium hydride
CaH2
UN1404
42.1
90 to 95%
Odorless
600°C (decomposition)
7789-78-8
MFCD00010897
14,1672
2mm and down
1.900
Moisture Sensitive
500g

Chemical Identifiers

7789-78-8
CaH2
MFCD00010897
Safety and Handling

Safety and Handling

GHS H Statement
H260;
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement
P231+P232-P280-P233-P370+P378c-P402+P404-P501a;
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: CALCIUM HYDRIDE

TSCA : TSCA

SDS Documents