Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium fluoride sputtering target, 12.7mm (0.5 in.) dia. x 2.5mm (0.098 in.) thick, 99.95% (metals basis)

Magnetron sputtering source

$2,219.64 - $2,219.64

Chemical Identifiers

Chemical Name or Material Calcium fluoride sputtering target
CAS 7789-75-5
Name Note 12.7mm (0.5 in.) dia. x 2.5mm (0.098 in.) thick
Molecular Formula CaF2
Formula Weight 78.08
MDL Number MFCD00010907
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43343KS

 
alfa aesar™
43343KS
1 each Each for $2,219.64
Description & Specifications

Specifications

Formula Weight 78.08
Chemical Name or Material Calcium fluoride sputtering target
TSCA TSCA
Odor Odorless
Name Note 12.7mm (0.5 in.) dia. x 2.5mm (0.098 in.) thick
Assay 99.95%
Assay Percent Notes (metals basis)
Molecular Formula CaF2
MDL Number MFCD00010907
CAS 7789-75-5
Quantity 1 each