Please login to your online account to display your discounted pricing

Calcium Fluoride (Precipitated Powder/Certified), Fisher Chemical


Quantity 250g
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLC89250

 View more versions of this product

Catalog No. C89-250

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 1422.8°C
Purity Grade Certified Grade
Formula Weight 78.07
Quantity 250g
Physical Form Solid
Color White
Density 3.180g/cm³
Ammonia 0.01% max.
Packaging Poly Bottle
CAS 7789-75-5
Heavy Metals (as Pb) 0.005% max.
ChemAlert Storage Symbol Gray
Chemical Name or Material Calcium Fluoride
MDL Number MFCD00010907
Identification Pass Test
Iron (Fe) 0.005% max.
Barium (Ba) 0.01% max.
Sulfate 0.05% max.
Chloride 0.05% max.
Molecular Formula CaF2

Specifications

Chemical Name or Material Calcium Fluoride
CAS 7789-75-5
Purity Grade Certified Grade
Physical Form Solid
Molecular Formula CaF2
Formula Weight 78.07
MDL Number MFCD00010907
Accessories
 

SDS & Safety Documents