Please login to your online account to display your discounted pricing

Calcium Fluoride (Precipitated Powder/Certified), Fisher Chemical

$316.46 - $529.46

Chemical Identifiers

Chemical Name or Material Calcium Fluoride
CAS 7789-75-5
Purity Grade Certified Grade
Physical Form Solid
Molecular Formula CaF2
Formula Weight 78.07
MDL Number MFCD00010907
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

C89-500

  SDS
fisher scientific company
FLC89500
Poly Bottle 500g Each for $529.46
Case of 6 Each for $3,176.76

C89-250

  SDS
fisher scientific company
FLC89250
Poly Bottle 250g Each for $316.46
Case of 6 Each for $1,898.76
Description & Specifications

Specifications

Chemical Name or Material Calcium Fluoride
CAS 7789-75-5
ChemAlert Storage Symbol Gray
Purity Grade Certified Grade
Physical Form Solid
Ammonia 0.01% max.
Molecular Formula CaF2
Formula Weight 78.07
MDL Number MFCD00010907
Barium (Ba) 0.01% max.
Chloride 0.05% max.
Density 3.180g/cm³
Heavy Metals (as Pb) 0.005% max.
Identification Pass Test
Iron (Fe) 0.005% max.
Packaging Poly Bottle
Sulfate 0.05% max.
Color White
Melting Point 1422.8°C
Quantity 500g
Safety and Handling