Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium fluoride crystal optic rectangle, 41mm x 23mm x 6mm, polished both sides

Chemical Identifiers

Chemical Name or Material Calcium fluoride crystal optic rectangle
Name Note 41mm x 23mm x 6mm, Polished both sides
MDL Number MFCD00010907
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38482KT

 
alfa aesar™
38482KT
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Calcium fluoride crystal optic rectangle
TSCA TSCA
Name Note 41mm x 23mm x 6mm, Polished both sides
MDL Number MFCD00010907
Quantity 1 each