Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium fluoride crystal optic rectangle, 38.5mm x 19.5mm x 4mm (drilled), polished both sides

Chemical Identifiers

Chemical Name or Material Calcium fluoride crystal optic rectangle
Name Note 38.5mm x 19.5mm x 4mm (drilled), Polished both sides
MDL Number MFCD00010907
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38476KT

 
alfa aesar™
38476KT
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1 each
Chemical Name or Material Calcium fluoride crystal optic rectangle
TSCA TSCA
Name Note 38.5mm x 19.5mm x 4mm (drilled), Polished both sides
MDL Number MFCD00010907