missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium fluoride crystal optic rectangle, 30mm x 15mm x 4mm, polished both sides

Spectroscopy, IR window

Chemical Identifiers

MDL Number MFCD00010907
Chemical Name or Material Calcium fluoride crystal optic rectangle, 30mm x 15mm x 4mm, Polished both sides
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA38458KT Alfa Aesar™
38458KT
1 each N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

MFCD00010907
Calcium fluoride crystal optic rectangle, 30mm x 15mm x 4mm, Polished both sides

Specifications

1 each
MFCD00010907
Calcium fluoride crystal optic rectangle, 30mm x 15mm x 4mm, Polished both sides

Safety and Handling

TSCA : TSCA