Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium fluoride crystal optic rectangle, 30mm x 15mm x 4mm (drilled), polished both sides

Chemical Identifiers

Chemical Name or Material Calcium fluoride crystal optic rectangle
Name Note 30mm x 15mm x 4mm (drilled), Polished both sides
MDL Number MFCD00010907
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38464KT

 
alfa aesar™
38464KT
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Name Note 30mm x 15mm x 4mm (drilled), Polished both sides
TSCA TSCA
Chemical Name or Material Calcium fluoride crystal optic rectangle
Quantity 1 each
MDL Number MFCD00010907