Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium fluoride crystal optic disc, 25mm x 2mm, polished both sides

Chemical Identifiers

Chemical Name or Material Calcium fluoride crystal optic disc
Name Note 25mm x 2mm, Polished both sides
MDL Number MFCD00010907
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38452KT

 
alfa aesar™
38452KT
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Name Note 25mm x 2mm, Polished both sides
MDL Number MFCD00010907
Chemical Name or Material Calcium fluoride crystal optic disc
Quantity 1 each