Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium fluoride crystal optic disc, 13mm x 1mm, polished both sides

Spectroscopy, IR window

Chemical Identifiers

Chemical Name or Material Calcium fluoride crystal optic disc
Name Note 13mm x 1mm, Polished both sides
MDL Number MFCD00010907
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38446KT

 
alfa aesar™
38446KT
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Calcium fluoride crystal optic disc
TSCA TSCA
Name Note 13mm x 1mm, Polished both sides
MDL Number MFCD00010907
Quantity 1 each