Learn More
Calcium fluoride, 99.5% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 011055A7
Description
Calcium fluoride is the commercially important source of fluoride compounds as well as hydrogen fluoride. It is transparent from ultraviolet to infrared region. It is used in spectroscopy, thermal imaging systems and excimer lasers. It is also used in the manufacturing of optical components including windows, prisms and lenses. In addition, it is also used in aluminum-metallurgy, brake lining manufacturing industries, and in tooth enamel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
ca 2500°C | |
1403°C | |
White | |
5 kg | |
CaF2 | |
EM>5, | |
Slightly soluble in acid. Insoluble in acetone. | |
[F-].[F-].[Ca++] | |
78.08 | |
CHEBI:35437 | |
Odorless | |
Hygroscopic | |
3.18 |
Calcium fluoride | |
7789-75-5 | |
Powder | |
99.5% (Metal basis) | |
MFCD00010907 | |
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade | |
WUKWITHWXAAZEY-UHFFFAOYSA-L | |
calcium difluoride | |
84512 | |
78.08 | |
1.434 | |
White powder |
Chemical Identifiers
7789-75-5 | |
78.08 | |
WUKWITHWXAAZEY-UHFFFAOYSA-L | |
84512 | |
[F-].[F-].[Ca++] |
CaF2 | |
MFCD00010907 | |
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade | |
CHEBI:35437 |
Safety and Handling
H315-H319-H335-H303
EINECSNumber : 232-188-7
RTECSNumber : EW1760000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only