missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium fluoride, 99%

Manufacturer:  Alfa Aesar™ 1102218

 View more versions of this product

Catalog No. AA1102218



Specifications

Specifications

Calcium fluoride
CaF2
14,1667
Soluble in ammonium salts. Insoluble in water.
[F-].[F-].[Ca+2]
78.075
CHEBI:35437
Transparent Crystals
3.18 g/mL
1.434
~2500°C
1403°C
7789-75-5
MFCD00010907
acmc-1bk0c, calcium 2+ difluoride, calcium fluoride, calcium fluoride 100g, calcium fluoride,optical grade, kalziumdifluorid, kalziumfluorid, ksc377c5h
WUKWITHWXAAZEY-UHFFFAOYSA-L
calcium;difluoride
84512
78.08
99%
Odorless
Hygroscopic
Undesignated
50g

Chemical Identifiers

7789-75-5
78.075
WUKWITHWXAAZEY-UHFFFAOYSA-L
84512
calcium;difluoride
CaF2
MFCD00010907
acmc-1bk0c, calcium 2+ difluoride, calcium fluoride, calcium fluoride 100g, calcium fluoride,optical grade, kalziumdifluorid, kalziumfluorid, ksc377c5h
CHEBI:35437
[F-].[F-].[Ca+2]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 232-188-7

RTECSNumber : EW1760000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents