missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium dihydrogen phosphate hydrate, 97%

Manufacturer:  Alfa Aesar™ A121230E

 View more versions of this product

Catalog No. AAA121230E



Description

Description

Applications
Calcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.

Solubility
Soluble in water 18 g/L @ 30°C.

Notes
Loses H{2}O at 100°, dec. at 200°
Specifications

Specifications

Calcium dihydrogen phosphate hydrate
Ca(H2PO4)2·xH2O
14,1693
234.06 (Anhydrous)
~200°C (decomposition)
301524-28-7
MFCD00010898
Calcium phosphate, monobasic; Calcium tetrahydrogen diphosphate
97%
2500g

Chemical Identifiers

301524-28-7
Ca(H2PO4)2·xH2O
MFCD00010898
Calcium phosphate, monobasic; Calcium tetrahydrogen diphosphate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TB8527000

TSCA : TSCA

SDS Documents