missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium D-gluconate gel, 2.5% w/w aq. soln.

Manufacturer:  Alfa Aesar™ 44542TJ

 View more versions of this product

Catalog No. AA44542TJ



Specifications

Specifications

Calcium D-gluconate gel
299-28-5
MFCD00064209
2.5% w/w aq. soln.
NEEHYRZPVYRGPP-IYEMJOQQSA-L
calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
9290
Gel
60g
C12H22CaO14
calcicol
Slowly soluble in water. Insoluble in alcohol,acetic acid and organic solvents.
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]
430.372
430.37

Chemical Identifiers

299-28-5
430.372
NEEHYRZPVYRGPP-IYEMJOQQSA-L
9290
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]
C12H22CaO14
MFCD00064209
calcicol
calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents