missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium cyanamide, tech.

Manufacturer:  Alfa Aesar™ 8946236

 View more versions of this product

Catalog No. AA8946236



Specifications

Specifications

Calcium cyanamide
CCaN2
MFCD00064894
4124391
80.11
Technical
Specific type
Gray
500g
156-62-7
CaNCN
UN1403
14,1662
Powder
2.29g/mL
Moisture Sensitive
1340°C

Chemical Identifiers

156-62-7
CCaN2
MFCD00064894
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: CALCIUM CYANAMIDE

RTECSNumber : GS6000000

TSCA : TSCA

SDS Documents