Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium crystalline dendritic pieces, 99.98% (metals basis)

Chemical Identifiers

Chemical Name or Material Calcium crystalline dendritic pieces
CAS 7440-70-2
MDL Number MFCD00085314
UN Number UN1401
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1445706

  SDS
alfa aesar™
1445706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1445714

  SDS
alfa aesar™
1445714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Packaging Ampule
Storage Note 1
  • Ampouled under argon
Chemical Name or Material Calcium crystalline dendritic pieces
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: CALCIUM
Assay 99.98%
Assay Percent Notes (metals basis)
TSCA TSCA
Health Hazard 3 GHS P Statement
P231+P232-P280-P233-P370+P378c-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H261
In contact with water releases flammable gas.
Odor Odorless
RTECS Number EV8040000
Physical Form Pieces
Health Hazard 1 Danger
CAS 7440-70-2
UN Number UN1401
Quantity 5g
MDL Number MFCD00085314

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P231+P232-P280-P233-P370+P378c-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H261
In contact with water releases flammable gas.

  • Ampouled under argon