missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium citrate tribasic tetrahydrate, 98+%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 390082500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium citrate tribasic tetrahydrate | |
10.0 % to 13.3 % (150°C, 4 hrs) | |
98.0 to 102% (on dried basis) (Complexometry) | |
C12H10Ca3O14·4H2O | |
250 g | |
calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f | |
LNIZKKFWMDARJV-UHFFFAOYSA-H | |
tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate | |
5282392 | |
98+% | |
0.2% max. (in HCl) |
5785-44-4 | |
Authentic | |
Plastic bottle | |
MFCD00150786 | |
15, 1663 | |
Solubility in water: 1g/L (20°C) | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2] | |
570.5 | |
570.5 | |
Analysis | |
Liquid |
Chemical Identifiers
5785-44-4 | |
570.5 | |
LNIZKKFWMDARJV-UHFFFAOYSA-H | |
5282392 | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2] |
C12H10Ca3O14·4H2O | |
MFCD00150786 | |
calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f | |
tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate |
RUO – Research Use Only