missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium citrate tetrahydrate, 96%

Manufacturer:  Alfa Aesar™ A148280E

 View more versions of this product

Catalog No. AAA148280E



Description

Description

Applications
Calcium citrate tetrahydrate may be used as raw material in the preparation of nanoparticle with the nominal composition Ca0.8 Ba0.2 Ti03 :Pr3+, via sol-gel process. It may be employed as carbon source in a simple and effective carbonization method to prepare porous carbon without any activation process. It is suitable for use in the preparation of bovine serum albumin (BSA) nanoparticles.

Solubility
Soluble in water and hydrochloric acid.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

Calcium citrate tetrahydrate
5785-44-4
MFCD00150786
LNIZKKFWMDARJV-UHFFFAOYSA-H
tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
5282392
96%
2500g
C12H18Ca3O18
calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, calcium citrate usp, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2]
570.492
572.60 (500.54 Anhydrous)

Chemical Identifiers

5785-44-4
570.492
LNIZKKFWMDARJV-UHFFFAOYSA-H
5282392
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2]
C12H18Ca3O18
MFCD00150786
calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, calcium citrate usp, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f
tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents