missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium chloride, White to gray powder, granules or beads, 99.99% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4428018

 View more versions of this product

Catalog No. AA4428018



Specifications

Specifications

Calcium chloride
99.99% (Metals basis)
MFCD00010903
Freely soluble in water,ethanol,methanol and acetic acid. Insoluble in liquid ammonia,dimethyl sulfoxide and ethyl acetate.
[Cl-].[Cl-].[Ca+2]
110.978
CHEBI:3312
Powder,Ampouled under argon
2.15
White to Gray
50g
10043-52-4
CaCl2
calcium chloride
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium;dichloride
5284359
110.99
Ultra dry
>1600°C
782°C

Chemical Identifiers

10043-52-4
110.978
UXVMQQNJUSDDNG-UHFFFAOYSA-L
5284359
calcium;dichloride
CaCl2
MFCD00010903
calcium chloride
CHEBI:3312
[Cl-].[Cl-].[Ca+2]
Safety and Handling

Safety and Handling

Do not breathe dust. Avoid contact with skin.

P264b-P280i-P305+P351+P338-P312-P501c

H319-H500

EINECSNumber : 233-140-8

RTECSNumber : EV9800000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents