Please login to your online account to display your discounted pricing

Calcium Chloride USP/FCC Granular, Macron Fine Chemicals™


macron

Quantity 500g
Packaging Glass

Manufacturer: avantor performance  461604

 View more versions of this product

Catalog No. MK4616500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging Glass
Quantity 500g
Chemical Name or Material Calcium Chloride
Name Note Dihydrate
CAS 10035-04-8
Purity Grade USP/FCC
Physical Form Granular
Molecular Formula CaCl2·2H2O
Formula Weight 147.01g/mol

Specifications

Chemical Name or Material Calcium Chloride
Name Note Dihydrate
CAS 10035-04-8
Purity Grade USP/FCC
Physical Form Granular
Molecular Formula CaCl2·2H2O
Formula Weight 147.01g/mol