Please login to your online account to display your discounted pricing

Calcium Chloride USP/FCC Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Calcium Chloride
Name Note Dihydrate
CAS 10035-04-8
Purity Grade USP/FCC
Physical Form Granular
Molecular Formula CaCl2·2H2O
Formula Weight 147.01g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK4616500

 
avantor performance
461604
Glass 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Packaging Glass
Quantity 500g
Chemical Name or Material Calcium Chloride
Name Note Dihydrate
CAS 10035-04-8
Purity Grade USP/FCC
Physical Form Granular
Molecular Formula CaCl2·2H2O
Formula Weight 147.01g/mol