Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium chloride, ultra dry, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Calcium chloride
CAS 10043-52-4
Name Note Ultra dry
Formula Weight 110.99
MDL Number MFCD00010903
Merck Index 14,1659
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3568609

  SDS
alfa aesar™
3568609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3568603

  SDS
alfa aesar™
3568603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3568618

  SDS
alfa aesar™
3568618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 110.99
Packaging Ampule
Chemical Name or Material Calcium chloride
TSCA TSCA
Mesh Size −10 Mesh
Name Note Ultra dry
Physical Form Beads
Assay 99.99%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00010903
CAS 10043-52-4
Merck Index 14,1659
Boiling Point >1600°C
Color White
Melting Point 782°C
Quantity 10g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Storage Note 1
  • Ampouled under argon
Density 2.15g/mL
RTECS Number EV9800000

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.

  • Ampouled under argon