missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium Chloride Standard, 400ppm Ca, Ricca Chemical

$56.06 - $83.91

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.086
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium;carbonate
SMILES C(=O)([O-])[O-].[Ca+2]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
1780-16 View Documents Ricca Chemical Company
RI178016
500mL Poly Bottle Each for $83.91
17804 SDS View Documents Ricca Chemical Company
RI17804
120mL Poly Bottle Each for $56.06
1780-32 SDS View Documents Ricca Chemical Company
RI178032
1L Poly Bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R17800004A View Documents Ricca Chemical Company
RIR17800004A
4L Poly natural N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

471-34-1
100.086
10112
calcium;carbonate
CCaO3
VTYYLEPIZMXCLO-UHFFFAOYSA-L
CHEBI:3311
C(=O)([O-])[O-].[Ca+2]

Specifications

471-34-1,7647-01-0,7732-18-5
100.0,0.2,0.1
1000ppm CaCO3 (400ppm Ca), 0.0100 M Ca2+, 0.0200 N Ca2+
C(=O)([O-])[O-].[Ca+2]
100.086
CHEBI:3311
1mL= 1 mg CaCO3
Calcium Chloride Standard
98.0,0.19,0.1
CCaO3
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium;carbonate
10112
Poly Bottle
500mL