missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium Chloride Standard, 1000 ppm CaCO3 (400 ppm Ca), 0.0100 M Ca2+, 0.0200 N Ca2+, Ricca Chemical
Supplier: Ricca Chemical Company 178016
Specifications
Calcium Chloride Standard | |
Natural Poly Bottle | |
98.0,0.19,0.1 | |
CCaO3 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
calcium carbonate | |
10112 | |
1000ppm CaCO3 (400ppm Ca), 0.0100 M Ca2+, 0.0200 N Ca2+ |
471-34-1,7647-01-0,7732-18-5 | |
500 mL | |
100.0,0.2,0.1 | |
MFCD00010906 | |
[Ca++].[O-]C([O-])=O | |
100.09 | |
CHEBI:3311 | |
1mL= 1 mg CaCO3 |
Chemical Identifiers
471-34-1 | |
100.09 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
CHEBI:3311 | |
[Ca++].[O-]C([O-])=O |
CCaO3 | |
MFCD00010906 | |
10112 | |
calcium carbonate |