Please login to your online account to display your discounted pricing

Calcium Chloride, Pellets, Purified, J.T.Baker™

For Desiccant Use

$213.17 - $700.28

Chemical Identifiers

Chemical Name or Material Calcium Chloride
CAS 10043-52-4
Purity Grade Purified
Molecular Formula CaCl2
Formula Weight 110.98
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002524

 
avantor performance
131305
Poly Bottle 2.5kg Each for $700.28

02002525

 
avantor performance
131301
Poly Bottle 500g Each for $213.17
Description & Specifications

Specifications

Physical Form Pellet
Formula Weight 110.98
Chemical Name or Material Calcium Chloride
Packaging Poly Bottle
Purity Grade Purified
Specific Gravity 1L = 2.15kg
Molecular Formula CaCl2
CAS 10043-52-4
Quantity 2.5kg