Learn More
Calcium chloride hydrate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01068018
Description
It is used as a food additive, as an electrolyte, it is used as a buffer in pools, an additive in plastics and in fire extinguishers. It is used as a fabric softener, dust control on roads. Calcium chloride is a soluble compound that can serve as a source of calcium ions in a solution. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium chloride hydrate | |
Crystalline Aggregates | |
(metals basis) | |
MFCD00149616 | |
calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g | |
QHFQAJHNDKBRBO-UHFFFAOYSA-L | |
calcium;dichloride;hexahydrate | |
6093252 | |
99.9965% | |
Odorless | |
1.71 g/mL |
22691-02-7 | |
50 g | |
CaCl2H12O6 | |
14,1659 | |
Soluble in water (5360mg/ml at 20°C),and alcohol. | |
O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2] | |
219.068 | |
110.99 (Anhydrous) | |
Puratronic™ | |
Hygroscopic |
Chemical Identifiers
22691-02-7 | |
219.068 | |
QHFQAJHNDKBRBO-UHFFFAOYSA-L | |
6093252 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2] |
CaCl2H12O6 | |
MFCD00149616 | |
calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g | |
calcium;dichloride;hexahydrate |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 607-129-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only