Please login to your online account to display your discounted pricing

Calcium chloride hexahydrate, extra pure, ACROS Organics™


Quantity 1kg
Packaging Plastic bottle

Manufacturer: fisher scientific acros  AC389250010

 View more versions of this product

Catalog No. AC389250010

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Extra Pure
Physical Form Chunks
Formula Weight 219.08
Quantity 1kg
Color Colorless
Merck Index 15, 1661
Assay 97.0 to 103.0%
Alkalinity 0.03% max. (as Ca(OH)2)
Chemical Name or Material Calcium chloride hexahydrate
Hydrochloric Acid 0.025% max.
Insoluble in Water 0.025% max.
Packaging Plastic bottle
Molecular Formula CaCl2·6H2O
Linear Formula CaCl2·6H2O
Sulfate 0.05% max.
CAS 7774-34-7
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Iron (Fe) 0.005% max.
Phosphate 0.003% max.
MDL Number MFCD00149616
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.

Specifications

Chemical Name or Material Calcium chloride hexahydrate
CAS 7774-34-7
Purity Grade Extra Pure
Assay 97.0 to 103.0%
Molecular Formula CaCl2·6H2O
Linear Formula CaCl2·6H2O
Formula Weight 219.08
MDL Number MFCD00149616
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents