Please login to your online account to display your discounted pricing

Calcium Chloride, Dihydrate, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$188.52 - $827.99

Chemical Identifiers

Chemical Name or Material Calcium Chloride
CAS 10035-04-8
Name Note Dihydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CaCl2·2H2O
Formula Weight 147.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002522

 
avantor performance
133201
Poly Bottle 500g Each for $188.52

02002526

 
avantor performance
133205
Poly Bottle 2.5kg Each for $827.99
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 147.01
Chemical Name or Material Calcium Chloride
Molecular Formula CaCl2·2H2O
CAS 10035-04-8
Name Note Dihydrate
Packaging Poly Bottle
Physical Form Granular
Specific Gravity 1L = 0.84kg
Quantity 500g